3-(3,4-Dihydroxyphenyl)-DL-alanine 96%
Catalog # APOSOR480320-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:197.19 g/mol
Storage Temperature:Ambient
CAS Number:63-84-3
Formula:C₉H₁₁NO₄
MDL Number:MFCD00063060