Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Dimethoxy-L-phenylalanine
3,4-Dimethoxy-L-phenylalanine 95%
3,4-Dimethoxy-L-phenylalanine 95%
3,4-Dimethoxy-L-phenylalanine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
3,4-Dimethoxy-L-phenylalanine 95%
Supplier:
Apollo Scientific
Certificates
About this item
CAS Number:
32161-30-1
:
Formula:
C₁₁H₁₅NO₄
MDL Number:
MFCD01321274
:
225.24 g/mol
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
Documentation