
2,3-Dimethyl-4-methoxybenzoic acid
Catalog # APOSOR47850-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:180.21 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:5628-61-5
Formula:C₁₀H₁₂O₃
MDL Number:MFCD06203271