

2,4,6-Collidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:108-75-8
:
Formula:C₈H₁₁N
MDL Number:MFCD00006338
Boiling Pt:171…172 °C
Melting Pt:–43 °C
Density:0.917 g/cm³ (25 °C)
Flash Pt:55 °C
Storage Temperature:Refrigerator
EINECS:203-613-3
ADR:3+6.1,III
:121.18 g/mol
UN:1992
:GHS02
:GHS06