
4,6-Dimethylnicotinic acid hydrochloride
Catalog # APOSOR4777-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:187.63 g/mol
Melting Pt:193…194 °C
:GHS07
Storage Temperature:Ambient
CAS Number:22047-86-5
Formula:C₈H₁₀ClNO₂
MDL Number:MFCD19103652