Order Entry
Export
ContactUsLinkComponent
2-Bromo-3,5-dichloropyridine
Irritant
2-Bromo-3,5-dichloropyridine
Catalog # APOSOR4756-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Bromo-3,5-dichloropyridine
Catalog # APOSOR4756-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :226.89 g/mol
Melting Pt:43 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:14482-51-0
Formula:C₅H₂BrCl₂N
MDL Number:MFCD00233990