Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Indandione 95%

1,3-Indandione 95%

Irritant
New Product
Supplier:   Apollo Scientific
1,3-Indandione 95%
Supplier:  Apollo Scientific
New Product
Certificates
1,3-Indandione 95%

5 Options Available Below

Product Details & Documents
Synonyms:  1,3-Indanedione, Indan-1,3-dione, 1H-Indene-1,3(2H)-dione
  • CAS Number:606-23-5
  • Formula:C₉H₆O₂
  • MDL Number:MFCD00003779
  • Molecular Weight:146.15 g/mol
  • Melting Pt:129…132 °C
  • Density:1.37 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:210-109-7
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 24,60 €
    24,60 €
    1 * 5 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 57,90 €
    57,90 €
    1 * 25 g
       57,90 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 228,00 €
    228,00 €
    1 * 100 g
       228,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1 130,00 €
    1 130,00 €
    1 * 500 g
       1 130,00 €

    Specifications:
    New Product
    New Product