1,4-Dibenzyloxy-2-nitrobenzene
Catalog # APOSOR471569-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:335.3500 g/mol
Storage Temperature:Ambient
CAS Number:51792-85-9
Formula:C₂₀H₁₇NO₄
MDL Number:MFCD00082778