Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-1,3-dinitrobenzene
2-Chloro-1,3-dinitrobenzene
2-Chloro-1,3-dinitrobenzene
2-Chloro-1,3-dinitrobenzene
Supplier:
Apollo Scientific
Certificates
Expand image
2-Chloro-1,3-dinitrobenzene
Supplier:
Apollo Scientific
About this item
CAS Number:
606-21-3
:
Formula:
C₆H₃ClN₂O₄
MDL Number:
MFCD00014689
:
202.55 g/mol
Storage Temperature:
Refrigerator
UN:
3441
ADR:
6.1,II