Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dibromoiodobenzene
2,4-Dibromoiodobenzene
2,4-Dibromoiodobenzene
APOSOR471033-5G
:
Apollo Scientific
:
2,4-Dibromoiodobenzene
APOSOR471033-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
361.8000 g/mol
:
Ambient
:
19393-94-3
:
C₆H₃Br₂I
:
MFCD09753709