6-Methoxy[1,1'-biphenyl]-3-carbaldehyde
Catalog # APOSOR471005-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:212.25 g/mol
Storage Temperature:Ambient
CAS Number:258831-56-0
Formula:C₁₄H₁₂O₂
MDL Number:MFCD06247960