Order Entry
Export
ContactUsLinkComponent
2-(1,3-Benzoxazol-2-ylamino)-6-methylpyrimidin-4-ol
2-(1,3-Benzoxazol-2-ylamino)-6-methylpyrimidin-4-ol
Catalog # APOSOR470449-5G
Supplier:  Apollo Scientific
CAS Number:  
2-(1,3-Benzoxazol-2-ylamino)-6-methylpyrimidin-4-ol
Catalog # APOSOR470449-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :242.24 g/mol
Storage Temperature:Ambient
CAS Number:86328-33-8
Formula:C₁₂H₁₀N₄O₂
MDL Number:MFCD01948713