

2,6-Dimethoxyaniline 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2734-70-5
:
Formula:C₈H₁₁NO₂
MDL Number:MFCD00053934
:153.18 g/mol
Melting Pt:76…78 °C
UN:2811
ADR:6.1,III
:GHS07
:GHS08
Storage Temperature:Refrigerator