3-Chloro[1,2,4]triazolo[4,3-a]pyridine
Catalog # APOSOR46696-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:153.57 g/mol
Storage Temperature:Ambient
CAS Number:4922-74-1
Formula:C₆H₄ClN₃
MDL Number:MFCD09901818