Order Entry
Export
ContactUsLinkComponent
 
N-Methyl-N-[(4-methylpyridin-2-yl)methyl]amine
Catalog # APOSOR46642-1G
Supplier:  Apollo Scientific
CAS Number:  
N-Methyl-N-[(4-methylpyridin-2-yl)methyl]amine
Catalog # APOSOR46642-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :136.2 g/mol
Storage Temperature:Ambient
CAS Number:1060804-82-1
Formula:C₈H₁₂N₂
MDL Number:MFCD09800577