

2,2'-Dithiodianiline (thulium ionophore I) 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1141-88-4
:
Formula:C₁₂H₁₂N₂S₂
MDL Number:MFCD00007703
:248.37 g/mol
Melting Pt:91…95 °C
EINECS:214-529-1
:GHS05
:GHS07
Storage Temperature:Ambient