
2,2-Bis(4-glycidyloxyphenyl)propane, Technical Grade
Catalog # APOSOR46614-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:340.42 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1675-54-3
Formula:C₂₁H₂₄O₄
MDL Number:MFCD00080480