Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dibromo-4-methylpyridine
2,6-Dibromo-4-methylpyridine
2,6-Dibromo-4-methylpyridine
:
Apollo Scientific
2,6-Dibromo-4-methylpyridine
:
Apollo Scientific
:
73112-16-0
:
:
C₆H₅Br₂N
:
MFCD11036225
:
250.93 g/mol
:
74…79 °C
:
GHS07
:
Ambient