Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tricyclohexylphosphine
Tricyclohexylphosphine 97%
Tricyclohexylphosphine 97%
Tricyclohexylphosphine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Tricyclohexylphosphine 97%
Supplier:
Apollo Scientific
Certificates
About this item
CAS Number:
2622-14-2
:
Formula:
C₁₈H₃₃P
MDL Number:
MFCD00003853
:
280.43 g/mol
Storage Temperature:
Ambient
Specifications
Environmentally Preferable:
Specifications
Related Information
Documentation
Certificates
Search by Lot Number
Documentation