
2,2'-Dibromoacetophenone 97%
Catalog # APOSOR46231-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:277.9406 g/mol
Boiling Pt:90…92 °C/0.02mmHg
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
CAS Number:49851-55-0
Formula:C₈H₆Br₂O
MDL Number:MFCD00044660