Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Bromo-3,5-dimethoxybenzene
1-Bromo-3,5-dimethoxybenzene 97%
1-Bromo-3,5-dimethoxybenzene 97%
Irritant
1-Bromo-3,5-dimethoxybenzene 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1-Bromo-3,5-dimethoxybenzene 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
20469-65-2
:
Formula:
C₈H₉BrO₂
MDL Number:
MFCD06200685
Melting Pt:
62…67 °C
:
217.06 g/mol
:
GHS07
Storage Temperature:
Ambient