Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dichloro-6-methylbenzamide
2,4-Dichloro-6-methylbenzamide
2,4-Dichloro-6-methylbenzamide
Irritant
2,4-Dichloro-6-methylbenzamide
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,4-Dichloro-6-methylbenzamide
Supplier:
Apollo Scientific
About this item
CAS Number:
175278-27-0
:
Formula:
C₈H₇Cl₂NO
MDL Number:
MFCD00205142
Melting Pt:
167…169 °C
:
204.06 g/mol
:
GHS07
Storage Temperature:
Ambient