Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,6-Dimethylsalicylaldehyde
3,6-Dimethylsalicylaldehyde
3,6-Dimethylsalicylaldehyde
3,6-Dimethylsalicylaldehyde
APOSOR46065-250MG
:
Apollo Scientific
:
3,6-Dimethylsalicylaldehyde
APOSOR46065-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
150.1700 g/mol
:
GHS07
:
Ambient
:
1666-04-2
:
C₉H₁₀O₂
:
MFCD04037328