Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Bromo-2H-benzo[b][1,4]oxazin-3(4H)-one
7-Bromo-2H-benzo[b][1,4]oxazin-3(4H)-one
7-Bromo-2H-benzo[b][1,4]oxazin-3(4H)-one
:
Apollo Scientific
7-Bromo-2H-benzo[b][1,4]oxazin-3(4H)-one
:
Apollo Scientific
:
321436-06-0
:
:
C₈H₆BrNO₂
:
MFCD07774199
:
228.0400 g/mol
:
GHS07
:
Ambient