
Methyl-2,4-dichloronicotinate
Catalog # APOSOR46030-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:206.022 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:442903-28-8
Formula:C₇H₅Cl₂NO₂
MDL Number:MFCD11100222