Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Chloro-2,2-dimethylpropane
1-Chloro-2,2-dimethylpropane
1-Chloro-2,2-dimethylpropane
:
Apollo Scientific
1-Chloro-2,2-dimethylpropane
:
Apollo Scientific
:
753-89-9
:
:
C₅H₁₁Cl
:
MFCD00000941
:
106.6 g/mol
:
84…85 °C
:
–20 °C
:
0.866 g/cm³ (25 °C)
:
–9 °C
:
Ambient
:
212-040-8
:
1993
:
3,II
:
GHS02
:
GHS07