2,2-Dichloropropane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:594-20-7
:
Formula:C₃H₆Cl₂
MDL Number:MFCD00000812
:112.99 g/mol
Boiling Pt:68…69 °C
Melting Pt:–35 °C
Density:1.07 g/cm³ (25 °C)
Flash Pt:–5 °C
Storage Temperature:Ambient
EINECS:209-832-0
UN:1993
ADR:3,II
:GHS02
:GHS07