Order Entry
Export
ContactUsLinkComponent
2,2-Dimethylpropane
Flammable
Compressed Gas
Environment
2,2-Dimethylpropane
Catalog # APOSOR460059-5G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Compressed Gas
Environment
2,2-Dimethylpropane
Catalog # APOSOR460059-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Cylinder
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :72.15 g/mol
Boiling Pt:9.5 °C
Density:0.613 g/cm³ (25 °C)
Flash Pt:–7.15 °C
Storage Temperature:Ambient
EINECS:207-343-7
UN:2044
ADR:2.1,
 :GHS02
 :GHS04
 :GHS09
CAS Number:463-82-1
Formula:C₅H₁₂