Order Entry
Export
ContactUsLinkComponent
Tetrachloroethylene 99%
Irritant
Health
Environment
Tetrachloroethylene 99%
Catalog # APOSOR460052-1KG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Health
Environment
Tetrachloroethylene 99%
Catalog # APOSOR460052-1KG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :165.83 g/mol
Boiling Pt:121 °C
Melting Pt:–22 °C
Density:1.623 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:204-825-9
UN:1897
ADR:6.1,III
 :GHS07
 :GHS08
 :GHS09
CAS Number:127-18-4
Formula:C₂Cl₄
MDL Number:MFCD00000834