Order Entry
Export
Orders LinkContactUsLinkComponent
1-Docosene

1-Docosene

Supplier:   Apollo Scientific
1-Docosene
Supplier:  Apollo Scientific
Certificates

2 Options Available Below

Product Details & Documents
  • CAS Number:1599-67-3
  • Formula:C₂₂H₄₄
  • MDL Number:MFCD00027085
  • Molecular Weight:308.59 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 151,00 €
    151,00 €
    1 * 25 g
       151,00 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 575,00 €
    575,00 €
    1 * 100 g
       575,00 €