4-(3,4-Methylenedioxyphenyl)-3-thiosemicarbazide
Catalog # APOSOR460017-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:211.24 g/mol
Storage Temperature:Ambient
CAS Number:206761-71-9
Formula:C₈H₉N₃O₂S
MDL Number:MFCD00060504