

(S)-(+)-3-Aminopiperidine dihydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:334618-07-4
:
Formula:C₅H₁₄Cl₂N₂
MDL Number:MFCD03427036
:173.08 g/mol
Melting Pt:205…209 °C
:GHS05
:GHS07
Storage Temperature:Ambient