Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(2,4-Dimethoxyphenyl)propionic acid
3-(2,4-Dimethoxyphenyl)propionic acid 97%
3-(2,4-Dimethoxyphenyl)propionic acid 97%
3-(2,4-Dimethoxyphenyl)propionic acid 97%
Supplier:
Apollo Scientific
Certificates
Expand image
3-(2,4-Dimethoxyphenyl)propionic acid 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
22174-29-4
:
Formula:
C₁₁H₁₄O₄
MDL Number:
MFCD00598205
:
210.23 g/mol
Storage Temperature:
Ambient