
(2-Bromo-4,5-dimethoxyphenyl)acetic acid
Catalog # APOSOR4518-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:275.1 g/mol
Melting Pt:94…96 °C
:GHS07
Storage Temperature:Ambient
CAS Number:4697-62-5
Formula:C₁₀H₁₁BrO₄
MDL Number:MFCD00016819