Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-1,4-dimethoxybenzene
2-Bromo-1,4-dimethoxybenzene
2-Bromo-1,4-dimethoxybenzene
:
Apollo Scientific
2-Bromo-1,4-dimethoxybenzene
:
Apollo Scientific
:
25245-34-5
:
:
C₈H₉BrO₂
:
MFCD00008355
:
217.06 g/mol
:
130…131 °C/10mmHg
:
1.45 g/cm³ (25 °C)
:
113 °C
:
246-756-7
:
GHS07
:
Ambient