Order Entry
Export
Orders LinkContactUsLinkComponent
1,9-Nonanedithiol 95+%

1,9-Nonanedithiol 95+%

Irritant
Supplier:   Apollo Scientific
1,9-Nonanedithiol 95+%
Supplier:  Apollo Scientific
Certificates
1,9-Nonanedithiol 95+%

3 Options Available Below

Product Details & Documents
Synonyms:  1,9-Dimercaptononane
  • CAS Number:3489-28-9
  • Formula:C₉H₂₀S₂
  • MDL Number:MFCD00004914
  • Molecular Weight:192.39 g/mol
  • Boiling Pt:284 °C
  • Density:0.952 g/cm³ (25 °C)
  • Flash Pt:102 °C
  • Storage Temperature:Ambient
  • UN:3334
  • ADR:9,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 42,00 €
    42,00 €
    1 * 5 g
       42,00 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 137,00 €
    137,00 €
    1 * 25 g
       137,00 €