Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Butanedithiol
1,4-Butanedithiol
1,4-Butanedithiol
Irritant
1,4-Butanedithiol
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,4-Butanedithiol
Supplier:
Apollo Scientific
About this item
CAS Number:
1191-08-8
:
Formula:
C₄H₁₀S₂
MDL Number:
MFCD00004906
:
122.26 g/mol
Boiling Pt:
105…106 °C
Density:
1.042 g/cm³ (25 °C)
:
GHS07
Storage Temperature:
Refrigerator