Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dibromoanisole
2,4-Dibromoanisole
2,4-Dibromoanisole
2,4-Dibromoanisole
Supplier:
Apollo Scientific
Certificates
Expand image
2,4-Dibromoanisole
Supplier:
Apollo Scientific
About this item
CAS Number:
21702-84-1
:
Formula:
C₇H₆Br₂O
MDL Number:
MFCD00000079
:
265.93 g/mol
Boiling Pt:
82…88 °C/0.3mmHg
Melting Pt:
60…64 °C
:
GHS07
Storage Temperature:
Ambient