3-(3,4-Dichlorophenoxy)azetidine hydrochloride
Catalog # APOSOR451012-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:254.55 g/mol
Storage Temperature:Ambient
CAS Number:606129-60-6
Formula:C₉H₁₀Cl₃NO
MDL Number:MFCD09264481