Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Diiodopropane

1,3-Diiodopropane

Irritant
Supplier:   Apollo Scientific
1,3-Diiodopropane
Supplier:  Apollo Scientific
Certificates
1,3-Diiodopropane

2 Options Available Below

Product Details & Documents
Synonyms:  Trimethylene diiodide
  • CAS Number:627-31-6
  • Formula:C₃H₆I₂
  • MDL Number:MFCD00001097
  • Molecular Weight:295.89 g/mol
  • Boiling Pt:111…113 °C/23mmHg
  • Melting Pt:–20 °C
  • Density:2.576 g/cm³ (25 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Refrigerator
  • EINECS:210-993-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 24,60 €
    24,60 €
    1 * 25 g
       24,60 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 79,60 €
    79,60 €
    1 * 100 g
       79,60 €