1,6-Dibromohexane 97+%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: Hexamethylen dibromide
- CAS Number:629-03-8
- Formula:BrH₂C(CH₂)₄CH₂Br
- MDL Number:MFCD00000272
- Molecular Weight:243.97 g/mol
- Boiling Pt:242…244 °C
- Melting Pt:–2 °C
- Density:1.586 g/cm³ (25 °C)
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:211-067-2
- UN:3082
- ADR:9+EHS,III
3 Options Available Below
Product Details & Documents
Synonyms: Hexamethylen dibromide
- CAS Number:629-03-8
- Formula:BrH₂C(CH₂)₄CH₂Br
- MDL Number:MFCD00000272
- Molecular Weight:243.97 g/mol
- Boiling Pt:242…244 °C
- Melting Pt:–2 °C
- Density:1.586 g/cm³ (25 °C)
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:211-067-2
- UN:3082
- ADR:9+EHS,III



