2,3-Dibromopyridine
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:13534-89-9
- Formula:C₅H₃Br₂N
- MDL Number:MFCD00234014
- Molecular Weight:236.89 g/mol
- Boiling Pt:249…250 °C
- Melting Pt:56…60 °C
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
- CAS Number:13534-89-9
- Formula:C₅H₃Br₂N
- MDL Number:MFCD00234014
- Molecular Weight:236.89 g/mol
- Boiling Pt:249…250 °C
- Melting Pt:56…60 °C
- Storage Temperature:Ambient



