Order Entry
Export
Orders LinkContactUsLinkComponent
4-(Dimethylamino)benzaldehyde 98%

4-(Dimethylamino)benzaldehyde 98%

Bottle, 100 g
Catalog # APOSOR43669-100G
Irritant
Supplier:   Apollo Scientific
4-(Dimethylamino)benzaldehyde 98%
Catalog # APOSOR43669-100G
Supplier:  Apollo Scientific
CAS Number:  100-10-7
4-(Dimethylamino)benzaldehyde 98%

Bottle, 100 g

CAS Number:  100-10-7
Price
31,90 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde
  • CAS Number:100-10-7
  • Formula:(CH₃)₂NC₆H₄CHO
  • MDL Number:MFCD00003381
  • Molecular Weight:149.19 g/mol
  • Boiling Pt:176…177 °C/17mmHg
  • Melting Pt:72…75 °C
  • Flash Pt:164 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications