
Ethyl-5-bromo-1,3,4-thiadiazole-2-carboxylate 97+%
Catalog # APOSOR43656-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:237.08 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1030613-07-0
Formula:C₅H₅BrN₂O₂S
MDL Number:MFCD12165919