Order Entry
Export
ContactUsLinkComponent
2,5-Dimethoxytetrahydrofuran 98%
Flammable
Irritant
2,5-Dimethoxytetrahydrofuran 98%
Catalog # APOSOR43635-500G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Irritant
2,5-Dimethoxytetrahydrofuran 98%
Catalog # APOSOR43635-500G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :132.16 g/mol
Boiling Pt:145…148 °C
Melting Pt:–45 °C
Density:1.021 g/cm³ (25 °C)
Flash Pt:35 °C
Storage Temperature:Ambient
EINECS:211-797-1
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:696-59-3
Formula:C₆H₁₂O₃
MDL Number:MFCD00005359