Order Entry
Export
ContactUsLinkComponent
4,5-Dimethyl-2-nitrophenol
Irritant
4,5-Dimethyl-2-nitrophenol
Catalog # APOSOR43630-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4,5-Dimethyl-2-nitrophenol
Catalog # APOSOR43630-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :167.17 g/mol
Melting Pt:80…82 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:18087-10-0
Formula:C₈H₉NO₃
MDL Number:MFCD15527214