Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Diacetylpyridine
2,6-Diacetylpyridine
2,6-Diacetylpyridine
Irritant
2,6-Diacetylpyridine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,6-Diacetylpyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
1129-30-2
:
Formula:
C₉H₉NO₂
MDL Number:
MFCD00006304
:
163.18 g/mol
Melting Pt:
79…82 °C
:
GHS07
Storage Temperature:
Ambient