
Methyl-3-hydroxy-2,2-dimethylpropanoate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14002-80-3
:
Formula:C₆H₁₂O₃
MDL Number:MFCD00009707
:132.1600 g/mol
Boiling Pt:177…178 °C
Melting Pt:–16 °C
Density:1.036 g/cm³ (25 °C)
Flash Pt:76 °C
EINECS:237-805-3
:GHS07
Storage Temperature:Ambient