
1,4-Diiodobenzene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:624-38-4
:
Formula:C₆H₄I₂
MDL Number:MFCD00001054
:329.91 g/mol
Boiling Pt:285 °C
Melting Pt:131…133 °C
Density:2.35 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:210-842-2
:GHS07