Order Entry
Export
Orders LinkContactUsLinkComponent
 

Benzophenone imine 97%

Irritant
Supplier:   Apollo Scientific
Benzophenone imine 97%
Supplier:  Apollo Scientific
Certificates
Benzophenone imine 97%

3 Options Available Below

Product Details & Documents
Synonyms:  Benzophenoneimine
  • CAS Number:1013-88-3
  • Formula:C₁₃H₁₁N
  • MDL Number:MFCD00001760
  • Molecular Weight:181.24 g/mol
  • Boiling Pt:151…153 °C/10mmHg
  • Density:1.08 g/cm³ (25 °C)
  • Flash Pt:109 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 24,60 €
    24,60 €
    1 * 25 g
       24,60 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 62,30 €
    62,30 €
    1 * 100 g
       62,30 €

  • Bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 197,00 €
    197,00 €
    1 * 500 g
       197,00 €